Assay Design



Gene Transcripts

Gene Symbol
HRH2
Entrez Gene ID
695226
Gene Name
histamine receptor H2
Gene Aliases
-
Chromosome Location
Chr.6: 173083262 - 173136431 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198697.3 NP_001185626.1 - 2782 101
GenBank mRNA JV565005.1 - - 1155 101

View Full Product Details