Assay Design



Gene Transcripts

Gene Symbol
GPR87
Entrez Gene ID
709766
Gene Name
G protein-coupled receptor 87
Gene Aliases
-
Chromosome Location
Chr.2: 53454723 - 53477608 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001107153.4 XP_001107153.1 - 508 140
XM_028843779.1 XP_028699612.1 - 631 140

View Full Product Details