Assay Design



Gene Transcripts

Gene Symbol
OR12D3
Entrez Gene ID
710321
Gene Name
olfactory receptor, family 12, subfamily D, member 3
Gene Aliases
-
Chromosome Location
Chr.4: 140814539 - 140815720 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194208.3 NP_001181137.3 - 946 137

View Full Product Details