Assay Design



Gene Transcripts

Gene Symbol
CGB8
Entrez Gene ID
574324
Gene Name
chorionic gonadotropin, beta polypeptide 8
Gene Aliases
CGB, hCGB
Chromosome Location
Chr.19: 49026385 - 49027549 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032928.1 NP_001028100.1 - 171 125
XM_015124374.2 XP_014979860.2 - 587 125
GenBank mRNA AY011015.1 - - 171 125

View Full Product Details