Assay Design



Gene Transcripts

Gene Symbol
MAP6
Entrez Gene ID
696223
Gene Name
microtubule associated protein 6
Gene Aliases
-
Chromosome Location
Chr.14: 68654488 - 68734770 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015115222.2 XP_014970708.2 - 1433 109

View Full Product Details