Assay Design



Gene Transcripts

Gene Symbol
ARL11
Entrez Gene ID
706725
Gene Name
ADP ribosylation factor like GTPase 11
Gene Aliases
-
Chromosome Location
Chr.17: 28085335 - 28088136 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194262.2 NP_001181191.2 - 128 160
GenBank mRNA JU473544.1 - - 169 160
JU473545.1 - - 169 160

View Full Product Details