Assay Design



Gene Transcripts

Gene Symbol
GJD3
Entrez Gene ID
700473
Gene Name
gap junction protein delta 3
Gene Aliases
-
Chromosome Location
Chr.16: 51516429 - 51521131 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015119587.2 XP_014975073.1 - 2185 127

View Full Product Details