Assay Design



Gene Transcripts

Gene Symbol
SP7
Entrez Gene ID
702575
Gene Name
Sp7 transcription factor
Gene Aliases
EGK_03709
Chromosome Location
Chr.11: 52897197 - 52909600 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001104653.3 XP_001104653.1 - 2882 86
XM_028829518.1 XP_028685351.1 - 116 86

View Full Product Details