Assay Design



Gene Transcripts

Gene Symbol
TAS2R38
Entrez Gene ID
695549
Gene Name
taste 2 receptor member 38
Gene Aliases
Mamu-T2R38
Chromosome Location
Chr.3: 168321356 - 168324277 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001085362.4 XP_001085362.3 - 1272 153

View Full Product Details