Assay Design



Gene Transcripts

Gene Symbol
CYP2D6
Entrez Gene ID
678691
Gene Name
cytochrome P450, family 2, subfamily D, polypeptide 6
Gene Aliases
CYP2D17, CYP2D42
Chromosome Location
Chr.10: 9363007 - 9367184 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040218.1 NP_001035308.1 - 842 146
GenBank mRNA AY635464.1 - - 842 146
GU981763.1 - - 850 146
JV708008.1 - - 499 146

View Full Product Details