Assay Design



Gene Transcripts

Gene Symbol
FBP1
Entrez Gene ID
709022
Gene Name
fructose-bisphosphatase 1
Gene Aliases
-
Chromosome Location
Chr.15: 113196598 - 113230208 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193652.1 NP_001180581.1 - 435 145
XM_015118081.2 XP_014973567.1 - 710 145
GenBank mRNA JV645221.1 - - 374 145

View Full Product Details