Assay Design



Gene Transcripts

Gene Symbol
PON3
Entrez Gene ID
699236
Gene Name
paraoxonase 3
Gene Aliases
EGK_13923
Chromosome Location
Chr.3: 110735309 - 110766243 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001095770.4 XP_001095770.1 - 728 146
XM_015134298.2 XP_014989784.1 - 784 146
GenBank mRNA JU325900.1 - - 651 146
JV063770.1 - - 694 146

View Full Product Details