Assay Design



Gene Transcripts

Gene Symbol
AFTPH
Entrez Gene ID
696100
Gene Name
aftiphilin
Gene Aliases
-
Chromosome Location
Chr.13: 44179616 - 44250292 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257727.1 NP_001244656.1 - 2505 111
XM_015112495.2 XP_014967981.1 - 2570 111
XM_015112496.2 XP_014967982.1 - 2325 111
GenBank mRNA JU332285.1 - - 2505 111
JU332286.1 - - 2505 111
JU472339.1 - - 2560 111
JU472340.1 - - 2560 111
JV044200.1 - - 2486 111
JV566312.1 - - 2569 111

View Full Product Details