Assay Design



Gene Transcripts

Gene Symbol
GPNMB
Entrez Gene ID
704990
Gene Name
glycoprotein nmb
Gene Aliases
-
Chromosome Location
Chr.3: 91829663 - 91903103 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266650.1 NP_001253579.1 - 300 135
XM_015133861.2 XP_014989347.1 - 453 135
XM_015133862.2 XP_014989348.1 - 542 135
XM_015133863.2 XP_014989349.1 - 454 135
XM_015133864.2 XP_014989350.1 - 243 135
XM_015133865.2 XP_014989351.1 - 522 135
GenBank mRNA JU332515.1 - - 320 135
JU473990.1 - - 300 135

View Full Product Details