Assay Design



Gene Transcripts

Gene Symbol
TMEM89
Entrez Gene ID
709488
Gene Name
transmembrane protein 89
Gene Aliases
EGK_11663
Chromosome Location
Chr.2: 104680575 - 104681474 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194540.2 NP_001181469.2 - 299 92

View Full Product Details