Assay Design



Gene Transcripts

Gene Symbol
ERMN
Entrez Gene ID
697560
Gene Name
ermin
Gene Aliases
-
Chromosome Location
Chr.12: 44412160 - 44419557 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001088029.4 XP_001088029.2 - 703 166
GenBank mRNA JV552058.1 - - 576 166

View Full Product Details