Assay Design



Gene Transcripts

Gene Symbol
ROPN1B
Entrez Gene ID
716215
Gene Name
rhophilin associated tail protein 1B
Gene Aliases
-
Chromosome Location
Chr.2: 150528520 - 150543697 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194626.3 NP_001181555.3 - 601 100

View Full Product Details