Assay Design



Gene Transcripts

Gene Symbol
THAP1
Entrez Gene ID
705845
Gene Name
THAP domain containing 1
Gene Aliases
-
Chromosome Location
Chr.8: 43452276 - 43459455 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257879.1 NP_001244808.1 - 1927 97
XM_015145248.2 XP_015000734.1 - 1759 97
XM_015145249.2 XP_015000735.1 - 1943 97
GenBank mRNA JU335241.1 - - 1927 97

View Full Product Details