Assay Design



Gene Transcripts

Gene Symbol
LRRN4CL
Entrez Gene ID
718662
Gene Name
LRRN4 C-terminal like
Gene Aliases
EGK_06103
Chromosome Location
Chr.14: 11509279 - 11511607 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194709.2 NP_001181638.1 - 439 73
GenBank mRNA JV054776.1 - - 166 73

View Full Product Details