Assay Design



Gene Transcripts

Gene Symbol
KRBA2
Entrez Gene ID
717353
Gene Name
KRAB-A domain containing 2
Gene Aliases
-
Chromosome Location
Chr.16: 8149649 - 8156824 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001113012.3 XP_001113012.3 - 899 117
XM_028836001.1 XP_028691834.1 - 97 117
GenBank mRNA JU325244.1 - - 82 117

View Full Product Details