Assay Design



Gene Transcripts

Gene Symbol
LOC696470
Entrez Gene ID
696470
Gene Name
dol-P-Glc:Glc(2)Man(9)GlcNAc(2)-PP-Dol alpha-1,2-glucosyltransferase
Gene Aliases
ALG10, ALG10B, EGK_03519
Chromosome Location
Chr.11: 35259474 - 35272556 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001086837.4 XP_001086837.1 - 506 108
GenBank mRNA JV707458.1 - - 389 108

View Full Product Details