Assay Design



Gene Transcripts

Gene Symbol
MAP3K19
Entrez Gene ID
712728
Gene Name
mitogen-activated protein kinase kinase kinase 19
Gene Aliases
YSK4
Chromosome Location
Chr.12: 20127909 - 20210973 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001101814.4 XP_001101814.4 - 1275 104

View Full Product Details