Assay Design



Gene Transcripts

Gene Symbol
LRRC66
Entrez Gene ID
698988
Gene Name
leucine rich repeat containing 66
Gene Aliases
-
Chromosome Location
Chr.5: 81631570 - 81663205 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001093042.4 XP_001093042.3 - 1812 104

View Full Product Details