Assay Design



Gene Transcripts

Gene Symbol
MARVELD2
Entrez Gene ID
700370
Gene Name
MARVEL domain containing 2
Gene Aliases
-
Chromosome Location
Chr.6: 67964254 - 67995152 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001094419.4 XP_001094419.1 - 1439 104
XM_015140247.2 XP_014995733.1 - 1857 104

View Full Product Details