Assay Design



Gene Transcripts

Gene Symbol
CDX1
Entrez Gene ID
711837
Gene Name
caudal type homeobox 1
Gene Aliases
-
Chromosome Location
Chr.6: 147680847 - 147698779 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001108012.4 XP_001108012.1 - 539 86

View Full Product Details