Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_015136681.2 | XP_014992167.1 | - | 1117 | 105 |
XM_015136688.2 | XP_014992174.1 | - | 1082 | 105 | |
XM_028835304.1 | XP_028691137.1 | - | 703 | 105 | |
GenBank mRNA | JU322260.1 | - | - | 651 | 105 |
JU474436.1 | - | - | 673 | 105 | |
JV046270.1 | - | - | 702 | 105 | |
JV643321.1 | - | - | 474 | 105 |