Assay Design



Gene Transcripts

Gene Symbol
DMTN
Entrez Gene ID
706762
Gene Name
dematin actin binding protein
Gene Aliases
EPB49
Chromosome Location
Chr.8: 22214344 - 22248806 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JU333339.1 - - 613 91
JV046423.1 - - 565 91
JV046424.1 - - 565 91
JV574083.1 - - 628 91
JV574084.1 - - 495 91
JV636019.1 - - 552 91
JV636020.1 - - 552 91

View Full Product Details