Assay Design



Gene Transcripts

Gene Symbol
LDHC
Entrez Gene ID
693731
Gene Name
lactate dehydrogenase C
Gene Aliases
EGK_06432
Chromosome Location
Chr.14: 47124215 - 47172388 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015114669.2 XP_014970155.2 - 584 126
XM_028833571.1 XP_028689404.1 - 569 126
XM_028833572.1 XP_028689405.1 - 451 126
XM_028833573.1 XP_028689406.1 - 563 126
XM_028833574.1 XP_028689407.1 - 569 126

View Full Product Details