Assay Design



Gene Transcripts

Gene Symbol
OCLN
Entrez Gene ID
700504
Gene Name
occludin
Gene Aliases
-
Chromosome Location
Chr.6: 68047516 - 68107491 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001094789.4 XP_001094789.1 - 1853 152
XM_001095022.4 XP_001095022.1 - 1627 152
XM_015140249.2 XP_014995735.1 - 1571 152
GenBank mRNA JU475761.1 - - 1523 152

View Full Product Details