Assay Design



Gene Transcripts

Gene Symbol
RAB3A
Entrez Gene ID
719400
Gene Name
RAB3A, member RAS oncogene family
Gene Aliases
EGK_10324
Chromosome Location
Chr.19: 17727276 - 17735118 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257767.1 NP_001244696.1 - 382 76
GenBank mRNA JU319824.1 - - 382 76
JU473388.1 - - 274 76
JV045780.1 - - 355 76

View Full Product Details