Assay Design



Gene Transcripts

Gene Symbol
CCL1
Entrez Gene ID
574177
Gene Name
C-C motif chemokine ligand 1
Gene Aliases
-
Chromosome Location
Chr.16: 30737365 - 30739968 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032849.1 NP_001028021.1 - 182 141
GenBank mRNA AF449264.1 - - 182 141

View Full Product Details