Assay Design



Gene Transcripts

Gene Symbol
TMBIM6
Entrez Gene ID
677702
Gene Name
transmembrane BAX inhibitor motif containing 6
Gene Aliases
TEGT
Chromosome Location
Chr.11: 49329287 - 49377732 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195449.1 NP_001182378.1 - 748 115
XM_015151592.2 XP_015007078.1 - 731 115
XM_015151593.2 XP_015007079.1 - 877 115
XM_015151595.2 XP_015007081.1 - 805 115
XM_015151597.2 XP_015007083.1 - 939 115
GenBank mRNA CB310349.1 - - 132 115
CN648400.1 - - 729 115
JV636703.1 - - 819 115

View Full Product Details