Assay Design



Gene Transcripts

Gene Symbol
URI1
Entrez Gene ID
700838
Gene Name
URI1 prefoldin like chaperone
Gene Aliases
C19H19orf2
Chromosome Location
Chr.19: 29372383 - 29464740 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001085459.4 XP_001085459.3 - 850 95
XM_015123477.2 XP_014978963.2 - 413 95
XM_015123478.2 XP_014978964.2 - 503 95
XM_015123480.2 XP_014978966.2 - 382 95
XM_015123483.2 XP_014978969.2 - 813 95
XM_028839928.1 XP_028695761.1 - 686 95
GenBank mRNA JV664417.1 - - 522 95
JV699930.1 - - 500 95

View Full Product Details