Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001266711.2 | NP_001253640.1 | - | 456 | 130 |
GenBank mRNA | JU321398.1 | - | - | 465 | 130 |
JU473147.1 | - | - | 472 | 130 | |
JV046863.1 | - | - | 456 | 130 | |
JV637252.1 | - | - | 475 | 130 |