Assay Design



Gene Transcripts

Gene Symbol
C5H4orf17
Entrez Gene ID
708639
Gene Name
chromosome 5 C4orf17 homolog
Gene Aliases
C4orf17
Chromosome Location
Chr.5: 97876003 - 97918641 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001107218.4 XP_001107218.2 - 764 119
XM_015138869.2 XP_014994355.1 - 864 119
XM_028848641.1 XP_028704474.1 - 676 119

View Full Product Details