Assay Design



Gene Transcripts

Gene Symbol
AJUBA
Entrez Gene ID
712865
Gene Name
ajuba LIM protein
Gene Aliases
EGK_18011, JUB
Chromosome Location
Chr.7: 84999875 - 85011797 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001104222.4 XP_001104222.1 - 1906 72
GenBank mRNA JV073934.1 - - 436 72
JV663941.1 - - 700 72

View Full Product Details