Assay Design



Gene Transcripts

Gene Symbol
SCGB3A2
Entrez Gene ID
709138
Gene Name
secretoglobin family 3A member 2
Gene Aliases
EGK_16993
Chromosome Location
Chr.6: 145204155 - 145294703 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001102976.4 XP_001102976.1 - 160 159

View Full Product Details