Assay Design



Gene Transcripts

Gene Symbol
SPSB3
Entrez Gene ID
722489
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
Gene Aliases
-
Chromosome Location
Chr.20: 1799783 - 1805548 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258151.1 NP_001245080.1 - 203 127
GenBank mRNA JU321201.1 - - 182 127
JU473292.1 - - 203 127
JV043616.1 - - 203 127
JV638609.1 - - 219 127

View Full Product Details