Assay Design



Gene Transcripts

Gene Symbol
HTR3C
Entrez Gene ID
706232
Gene Name
5-hydroxytryptamine receptor 3C
Gene Aliases
-
Chromosome Location
Chr.2: 89336488 - 89344196 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028843483.1 XP_028699316.1 - 929 135

View Full Product Details