Assay Design



Gene Transcripts

Gene Symbol
ESAM
Entrez Gene ID
712200
Gene Name
endothelial cell adhesion molecule
Gene Aliases
EGK_07047
Chromosome Location
Chr.14: 117531273 - 117540258 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257484.1 NP_001244413.1 - 190 89
GenBank mRNA JU337021.1 - - 190 89
JU473462.1 - - 228 89

View Full Product Details