Assay Design



Gene Transcripts

Gene Symbol
TRUB1
Entrez Gene ID
699133
Gene Name
TruB pseudouridine synthase family member 1
Gene Aliases
-
Chromosome Location
Chr.9: 115460021 - 115497975 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266998.2 NP_001253927.1 - 440 107
GenBank mRNA JU333034.1 - - 470 107
JU474380.1 - - 440 107

View Full Product Details