Assay Design



Gene Transcripts

Gene Symbol
WNT1
Entrez Gene ID
709009
Gene Name
Wnt family member 1
Gene Aliases
-
Chromosome Location
Chr.11: 48586494 - 48590772 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001105191.4 XP_001105191.1 - 669 103

View Full Product Details