Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258068.1 | NP_001244997.1 | - | 521 | 115 |
XM_015117912.2 | XP_014973398.1 | - | 517 | 115 | |
XM_028826638.1 | XP_028682471.1 | - | 517 | 115 | |
GenBank mRNA | JU472995.1 | - | - | 521 | 115 |