Assay Design



Gene Transcripts

Gene Symbol
MYF5
Entrez Gene ID
697426
Gene Name
myogenic factor 5
Gene Aliases
EGK_03968
Chromosome Location
Chr.11: 80246556 - 80249502 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001087187.4 XP_001087187.1 - 834 157

View Full Product Details