Assay Design



Gene Transcripts

Gene Symbol
CACNG2
Entrez Gene ID
696294
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 2
Gene Aliases
-
Chromosome Location
Chr.10: 14861193 - 14999487 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028827749.1 XP_028683582.1 - 1512 97
GenBank mRNA JU332485.1 - - 400 97
JV568084.1 - - 404 97
JV670446.1 - - 366 97

View Full Product Details