Assay Design



Gene Transcripts

Gene Symbol
SYPL1
Entrez Gene ID
698350
Gene Name
synaptophysin like 1
Gene Aliases
-
Chromosome Location
Chr.3: 132398468 - 132421154 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266817.1 NP_001253746.1 - 127 80
GenBank mRNA JU323179.1 - - 129 80
JU476961.1 - - 74 80
JV045068.1 - - 127 80

View Full Product Details