Assay Design



Gene Transcripts

Gene Symbol
RPP14
Entrez Gene ID
706788
Gene Name
ribonuclease P/MRP subunit p14
Gene Aliases
EGK_21389
Chromosome Location
Chr.2: 114457021 - 114468914 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266712.1 NP_001253641.1 - 453 100
XM_015130593.2 XP_014986079.1 - 747 100
XM_015130594.2 XP_014986080.1 - 518 100
XM_015130595.2 XP_014986081.1 - 413 100
XM_015130596.2 XP_014986082.1 - 343 100
XM_015130597.2 XP_014986083.1 - 750 100
XM_028843253.1 XP_028699086.1 - 560 100
XM_028843254.1 XP_028699087.1 - 537 100
GenBank mRNA JU332688.1 - - 410 100
JU472057.1 - - 283 100
JU472058.1 - - 627 100
JV044948.1 - - 453 100
JV044949.1 - - 420 100
JV044950.1 - - 453 100

View Full Product Details