Assay Design



Gene Transcripts

Gene Symbol
OIP5
Entrez Gene ID
705837
Gene Name
Opa interacting protein 5
Gene Aliases
EGK_17383
Chromosome Location
Chr.7: 17768015 - 17784052 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258125.1 NP_001245054.1 - 429 112
GenBank mRNA JU474204.1 - - 429 112

View Full Product Details