Assay Design



Gene Transcripts

Gene Symbol
FBXO22
Entrez Gene ID
707264
Gene Name
F-box protein 22
Gene Aliases
-
Chromosome Location
Chr.7: 53104112 - 53135717 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266238.2 NP_001253167.1 - 193 57
GenBank mRNA JU333946.1 - - 214 57
JU475131.1 - - 193 57
JV046730.1 - - 156 57

View Full Product Details