Assay Design



Gene Transcripts

Gene Symbol
VAX2
Entrez Gene ID
703531
Gene Name
ventral anterior homeobox 2
Gene Aliases
-
Chromosome Location
Chr.13: 37812366 - 37845935 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001100565.4 XP_001100565.3 - 326 74

View Full Product Details